missing translation for 'onlineSavingsMsg'
Learn More

Cefaclor, Thermo Scientific™

A second-generation cephalosporin antibiotic

£57.40 - £203.01

Chemical Identifiers

CAS 53994-73-3
Molecular Formula C15H14ClN3O4S
Molecular Weight (g/mol) 367.804
MDL Number MFCD00151471
InChI Key QYIYFLOTGYLRGG-GPCCPHFNSA-N
Synonym Panoral; (R)-3-Chloro-7-((S)-2-amino-2-phenylacetamido)-3-cephem-4-carboxylic acid
PubChem CID 51039
ChEBI CHEBI:3478
IUPAC Name (6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES C1C(=C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15406289
SDS View Documents
Thermo Scientific Alfa Aesar
J64252.03
1g
ONLINE EXCLUSIVE PRICE
£58.70
£57.40 / 1g
Save £1.30
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15416289
SDS View Documents
Thermo Scientific Alfa Aesar
J64252.09
10g
ONLINE EXCLUSIVE PRICE
£207.00
£203.01 / 10g
Save £3.99
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Cefaclor is used to study urinary tract, intra-abdominal, and Haemophilus influenzae infections. It is used to study the mechanism of human renal organic anion and peptide transporters such as hOAT1, hPEPT1, and hPEPT2 and to study the effects of inhibition of penicillin-binding proteins on bacterial cell wall mucopeptide synthesis.

Solubility
Soluble in 1N HCl

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

53994-73-3
367.804
QYIYFLOTGYLRGG-GPCCPHFNSA-N
51039
(6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
C15H14ClN3O4S
MFCD00151471
Panoral; (R)-3-Chloro-7-((S)-2-amino-2-phenylacetamido)-3-cephem-4-carboxylic acid
CHEBI:3478
C1C(=C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)Cl

Specifications

53994-73-3
C15H14ClN3O4S
8176092
Panoral; (R)-3-Chloro-7-((S)-2-amino-2-phenylacetamido)-3-cephem-4-carboxylic acid
QYIYFLOTGYLRGG-GPCCPHFNSA-N
(6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
51039
367.81
White to Yellow
MFCD00151471
14,1912
Soluble in 1N HCl
C1C(=C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)Cl
367.804
CHEBI:3478
Cefaclor
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c

H317-H334

missing translation for 'einecsNumber' : 258-909-5

missing translation for 'rtecsNumber' : XI0363000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.