missing translation for 'onlineSavingsMsg'
Learn More

Cefaclor, Thermo Scientific™

A second-generation cephalosporin antibiotic

Brand:  Thermo Scientific Alfa Aesar J64252.03

Quantity 1g

 View more versions of this product

Product Code. 15406289

  • ONLINE EXCLUSIVE PRICE
    £58.70
    £57.40 / 1g
    Save £1.30
    2% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Cefaclor is used to study urinary tract, intra-abdominal, and Haemophilus influenzae infections. It is used to study the mechanism of human renal organic anion and peptide transporters such as hOAT1, hPEPT1, and hPEPT2 and to study the effects of inhibition of penicillin-binding proteins on bacterial cell wall mucopeptide synthesis.

Solubility
Soluble in 1N HCl

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

53994-73-3
367.804
QYIYFLOTGYLRGG-GPCCPHFNSA-N
51039
(6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
C15H14ClN3O4S
MFCD00151471
Panoral; (R)-3-Chloro-7-((S)-2-amino-2-phenylacetamido)-3-cephem-4-carboxylic acid
CHEBI:3478
C1C(=C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)Cl

Specifications

53994-73-3
1g
MFCD00151471
14,1912
Soluble in 1N HCl
C1C(=C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)Cl
367.804
CHEBI:3478
Cefaclor
White to Yellow
C15H14ClN3O4S
8176092
Panoral; (R)-3-Chloro-7-((S)-2-amino-2-phenylacetamido)-3-cephem-4-carboxylic acid
QYIYFLOTGYLRGG-GPCCPHFNSA-N
(6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
51039
367.81
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.