missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CECR5 Recombinant Rabbit Monoclonal Antibody (23GB6585)

Rabbit Recombinant Monoclonal Antibody

£172.00 - £488.00

Specifications

Antigen CECR5
Clone 23GB6585
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30274791
View Documents
Invitrogen™
MA557422
100 μL
£488.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30276007
View Documents
Invitrogen™
MA557423
20 μL
£172.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CECR5 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

HDHD5 (Haloacid Dehalogenase Like Hydrolase Domain Containing 5) is a Protein Coding gene. Diseases associated with HDHD5 include Cat Eye Syndrome. An important paralog of this gene is CRLS1. [GeneCards]
Specifications

Specifications

CECR5
-20°C
Recombinant Monoclonal
Liquid
IgG
Human
HDHD5
A930002G03Rik; cat eye syndrome chromosome region, candidate 5; cat eye syndrome chromosome region, candidate 5 homolog; cat eye syndrome critical region protein 5; Cat eye syndrome critical region protein 5 homolog; CECR5; Haloacid dehalogenase-like hydrolase domain-containing 5; HDHD5
HDHD5
Primary
Affinity chromatography
23GB6585
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9BXW7
27440
A synthesized peptide derived from human CECR5 (aa 370-396).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.