missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CECR5 Recombinant Rabbit Monoclonal Antibody (23GB6585)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557423
This item is not returnable.
View return policy
Description
CECR5 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
HDHD5 (Haloacid Dehalogenase Like Hydrolase Domain Containing 5) is a Protein Coding gene. Diseases associated with HDHD5 include Cat Eye Syndrome. An important paralog of this gene is CRLS1. [GeneCards].
Specifications
| CECR5 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| HDHD5 | |
| A930002G03Rik; cat eye syndrome chromosome region, candidate 5; cat eye syndrome chromosome region, candidate 5 homolog; cat eye syndrome critical region protein 5; Cat eye syndrome critical region protein 5 homolog; CECR5; Haloacid dehalogenase-like hydrolase domain-containing 5; HDHD5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27440 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB6585 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BXW7 | |
| HDHD5 | |
| A synthesized peptide derived from human CECR5 (aa 370-396). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido