missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEBP Beta Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£158.00 - £348.00
Specifications
| Antigen | CEBP Beta |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18312945
|
Novus Biologicals
NBP3-16923-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18325774
|
Novus Biologicals
NBP3-16923-100UL |
100 μg |
£348.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CEBP Beta Polyclonal antibody specifically detects CEBP Beta in Human, Mouse samples. It is validated for Western BlotSpecifications
| CEBP Beta | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| C/EBP beta, C/EBP-beta, CCAAT/enhancer binding protein (C/EBP), beta, CCAAT/enhancer-binding protein beta, CRP2, IL6DBP, interleukin 6-dependent DNA-binding protein, LAPTCF-5, Liver activator protein, liver-enriched transcriptional activator protein, MGC32080, NF-IL6, Nuclear factor NF-IL6, nuclear factor of interleukin 6, TCF5NFIL6, Transcription factor 5 | |
| A synthetic phosphorylated peptide around T235 of human CEBP Beta (NP_005185.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific | |
| PBS, 50% glycerol, pH7.3 | |
| 1051 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title