missing translation for 'onlineSavingsMsg'
Learn More

CEBP Beta Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18312945 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18312945 20 μg 20µL
18325774 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18312945 Supplier Novus Biologicals Supplier No. NBP31692320UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CEBP Beta Polyclonal antibody specifically detects CEBP Beta in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEBP Beta
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias C/EBP beta, C/EBP-beta, CCAAT/enhancer binding protein (C/EBP), beta, CCAAT/enhancer-binding protein beta, CRP2, IL6DBP, interleukin 6-dependent DNA-binding protein, LAPTCF-5, Liver activator protein, liver-enriched transcriptional activator protein, MGC32080, NF-IL6, Nuclear factor NF-IL6, nuclear factor of interleukin 6, TCF5NFIL6, Transcription factor 5
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around T235 of human CEBP Beta (NP_005185.2).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 1051
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.