missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDYL2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£171.00 - £378.00
Specifications
| Antigen | CDYL2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18697942
|
Novus Biologicals
NBP2-92495-0.02ml |
0.02 mL |
£171.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18693102
|
Novus Biologicals
NBP2-92495-0.1ml |
0.1 mL |
£378.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDYL2 Polyclonal antibody specifically detects CDYL2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| CDYL2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| CDY-like 2, chromodomain protein, Y-like 2, chromodomain Y-like protein 2, FLJ38866 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-230 of human CDYL2 (NP_689555.2). MSKDKRIKSGKQSSTSKLLRDSRGPSVEKLSHRPSDPGKSKGTSHKRKRINPPLAKPKKGYSGKPSSGGDRATKTVSYRTTPSGLQIMPLKKSQNGMENGDAGSEKDERHFGNGSHQPGLDLNDHVGEQDMGECDVNHATLAENGLGSALTNGGLNLHSPVKRKLEAEKDY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 124359 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title