missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDYL2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92495-0.1ml
This item is not returnable.
View return policy
Description
CDYL2 Polyclonal antibody specifically detects CDYL2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CDYL2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CDY-like 2, chromodomain protein, Y-like 2, chromodomain Y-like protein 2, FLJ38866 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-230 of human CDYL2 (NP_689555.2). MSKDKRIKSGKQSSTSKLLRDSRGPSVEKLSHRPSDPGKSKGTSHKRKRINPPLAKPKKGYSGKPSSGGDRATKTVSYRTTPSGLQIMPLKKSQNGMENGDAGSEKDERHFGNGSHQPGLDLNDHVGEQDMGECDVNHATLAENGLGSALTNGGLNLHSPVKRKLEAEKDY | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 124359 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction