missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDX2 Mouse, Clone: CDX2/1690, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £406.00
Specifications
Antigen | CDX2 |
---|---|
Clone | CDX2/1690 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15969727
|
EnquireBio
1045-MSM1-P1 |
100 μg |
£406.00
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15959727
|
EnquireBio
1045-MSM1-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
CDX2 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
1045 | |
Recombinant human CDX2 protein fragment (aa150-249) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
40kDa |
CDX2/1690 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
XP_011533178 | |
CDX2 | |
IgG2a κ | |
Protein A/G Purified | |
Nuclear | |
HT29 cells or Colon Carcinoma |