missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDX2 Antibody (PCRP-CDX2-1A3), Novus Biologicals™
Mouse Monoclonal Antibody
£211.00 - £373.00
Specifications
| Antigen | CDX2 |
|---|---|
| Clone | PCRP-CDX2-1A3 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Microarray, Protein Array |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Microarray, Protein Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18603399
|
Novus Biologicals
NBP2-75758-100ug |
100 μg |
£373.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627386
|
Novus Biologicals
NBP2-75758-20ug |
20 μg |
£211.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDX2 Monoclonal antibody specifically detects CDX2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Microarray, Protein ArraySpecifications
| CDX2 | |
| 0.2 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Microarray, Protein Array | |
| Unconjugated | |
| Mouse | |
| Cancer, Chromatin Research, DNA Repair, Growth and Development, Transcription Factors and Regulators | |
| 10 mM PBS with 0.05% BSA | |
| caudal type homeo box transcription factor 2, caudal type homeobox 2, Caudal-type homeobox protein 2, CDX-3, CDX3caudal type homeobox transcription factor 2, homeobox protein CDX-2 | |
| Recombinant human full-length CDX2 protein (Uniprot: Q99626) | |
| Primary | |
| Store at 4°C. |
| PCRP-CDX2-1A3 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Microarray, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q99626, Q99626, Q99626, Q99626 | |
| 1045 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title