missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDX2 Antibody (PCRP-CDX2-1A3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-75758-20ug
This item is not returnable.
View return policy
Description
CDX2 Monoclonal antibody specifically detects CDX2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Microarray, Protein Array
Specifications
| CDX2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Microarray, Protein Array | |
| Q99626, Q99626, Q99626, Q99626 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Microarray, Protein Array | |
| PCRP-CDX2-1A3 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| caudal type homeo box transcription factor 2, caudal type homeobox 2, Caudal-type homeobox protein 2, CDX-3, CDX3caudal type homeobox transcription factor 2, homeobox protein CDX-2 | |
| Recombinant human full-length CDX2 protein (Uniprot: Q99626) | |
| 20 μg | |
| Cancer, Chromatin Research, DNA Repair, Growth and Development, Transcription Factors and Regulators | |
| 1045 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction