missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDT1 Recombinant Rabbit Monoclonal Antibody (23GB5790), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | CDT1 |
|---|---|
| Clone | 23GB5790 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30088810
|
Invitrogen™
MA553924 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089471
|
Invitrogen™
MA553925 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDT1 Recombinant Monoclonal antibody specifically detects CDT1 in Human samples. It is validated for Western BlotSpecifications
| CDT1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9H211 | |
| 81620 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5790 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2610318F11Rik; AW545653; C76791; CDT1; chromatin licensing and DNA replication factor 1; DNA replication factor Cdt1; Double parked homolog; Double parked, Drosophila, homolog of; DUP; Retroviral insertion site 2 protein; retroviral integration site 1; retroviral integration site 2; RIS2 | |
| A synthesized peptide derived from human CDT1 (230-546AA). | |
| Primary | |
| -20°C | |
| CDT1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title