missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDT1 Recombinant Rabbit Monoclonal Antibody (23GB5790), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553924
This item is not returnable.
View return policy
Description
KMT2A is a histone methyltransferase that plays an essential role in early development and hematopoiesis. It is a catalytic subunit of the MLL1/MLL complex, a multiprotein complex that mediates both methylation of 'Lys-4' of histone H3 (H3K4me) complex and acetylation of 'Lys-16' of histone H4 (H4K16ac). In the MLL1/MLL complex, KMT2A specifically mediates H3K4me, a tag for epigenetic transcriptional activation. KMT2A has weak methyltransferase activity by itself, and requires other components of the MLL1/MLL complex to obtain full methyltransferase activity. It has no activity toward histone H3 phosphorylated on 'Thr-3', less activity toward H3 dimethylated on 'Arg-8' or 'Lys-9', and has higher activity toward H3 acetylated on 'Lys-9'. Required for transcriptional activation of HOXA9 and promotes PPP1R15A-induced apoptosis.
Specifications
| CDT1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CDT1 | |
| 2610318F11Rik; AW545653; C76791; CDT1; chromatin licensing and DNA replication factor 1; DNA replication factor Cdt1; Double parked homolog; Double parked, Drosophila, homolog of; DUP; Retroviral insertion site 2 protein; retroviral integration site 1; retroviral integration site 2; RIS2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 81620 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5790 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9H211 | |
| CDT1 | |
| A synthesized peptide derived from human CDT1 (230-546AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction