missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDT1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £358.00
Specifications
| Antigen | CDT1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
CDT1 Polyclonal antibody specifically detects CDT1 in Mouse samples. It is validated for Western BlotSpecifications
| CDT1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cell Cycle and Replication | |
| PBS with 50% glycerol, pH7.3. | |
| 81620 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| chromatin licensing and DNA replication factor 1, DNA replication factor Cdt1, Double parked homolog, Double parked, Drosophila, homolog of, DUPRIS2 | |
| A synthetic peptide corresponding to a sequence within amino acids 400 to the C-terminus of human CDT1 (NP_112190.2). PATPPATPPAASPSALKGVSQDLLERIRAKEAQKQLAQMTRCPEQEQRLQRLERLPELARVLRSVFVSERKPALSMEVACARMVGSCCTIMSPGEMEKHLLLLSELLPDWLSLHRIRTDTYVKLDKAADLAHITARLAHQTRAEEGL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title