missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDT1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92328-0.1ml
This item is not returnable.
View return policy
Description
CDT1 Polyclonal antibody specifically detects CDT1 in Mouse samples. It is validated for Western Blot
Specifications
| CDT1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| chromatin licensing and DNA replication factor 1, DNA replication factor Cdt1, Double parked homolog, Double parked, Drosophila, homolog of, DUPRIS2 | |
| A synthetic peptide corresponding to a sequence within amino acids 400 to the C-terminus of human CDT1 (NP_112190.2). PATPPATPPAASPSALKGVSQDLLERIRAKEAQKQLAQMTRCPEQEQRLQRLERLPELARVLRSVFVSERKPALSMEVACARMVGSCCTIMSPGEMEKHLLLLSELLPDWLSLHRIRTDTYVKLDKAADLAHITARLAHQTRAEEGL | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication | |
| 81620 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction