missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£175.00 - £367.00
Specifications
| Antigen | CDR2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 ug/ml, Flow Cytometry reported by customer review, Immunohistochemistry 5-10 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunohistochemistry-Paraffin 5-10 ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18736253
|
Novus Biologicals
NB110-58345SS |
0.025 mL |
£175.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18798843
|
Novus Biologicals
NB110-58345 |
0.1 mL |
£367.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDR2 Polyclonal specifically detects CDR2 in Human, Mouse, Bovine, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| CDR2 | |
| Western Blot 2 ug/ml, Flow Cytometry reported by customer review, Immunohistochemistry 5-10 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunohistochemistry-Paraffin 5-10 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Cancer, Immune System Diseases, Immunology, Neuroscience | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide | |
| CDR62, cerebellar degeneration-related protein (62kD), cerebellar degeneration-related protein 2, cerebellar degeneration-related protein 2, 62kDa, Major Yo paraneoplastic antigen, Paraneoplastic cerebellar degeneration-associated antigen, PCD17, Yo, Yo paraneoplastic antigen | |
| CDR2 | |
| IgG | |
| Affinity Purified | |
| Human, mouse, bovine, primate. Immunogen has 85% homology to rat. |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Bovine, Primate | |
| Q01850 | |
| 1039 | |
| A synthetic peptide made to a C-terminal region of human CDR2 [Swiss-Prot# Q01850] | |
| Primary | |
| Store at 4C. Do not freeze. | |
| 51 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title