missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB110-58345SS
This item is not returnable.
View return policy
Description
CDR2 Polyclonal specifically detects CDR2 in Human, Mouse, Bovine, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CDR2 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide | |
| CDR62, cerebellar degeneration-related protein (62kD), cerebellar degeneration-related protein 2, cerebellar degeneration-related protein 2, 62kDa, Major Yo paraneoplastic antigen, Paraneoplastic cerebellar degeneration-associated antigen, PCD17, Yo, Yo paraneoplastic antigen | |
| Rabbit | |
| 51 kDa | |
| 0.025 mL | |
| Cancer, Immune System Diseases, Immunology, Neuroscience | |
| 1039 | |
| Human, Mouse, Bovine, Primate | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 2 ug/ml, Flow Cytometry reported by customer review, Immunohistochemistry 5-10 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunohistochemistry-Paraffin 5-10 ug/ml | |
| Q01850 | |
| CDR2 | |
| A synthetic peptide made to a C-terminal region of human CDR2 [Swiss-Prot# Q01850] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, mouse, bovine, primate. Immunogen has 85% homology to rat. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction