missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDKN2C Recombinant Rabbit Monoclonal Antibody (23GB5650), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | CDKN2C |
|---|---|
| Clone | 23GB5650 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089817
![]() |
Invitrogen™
MA552886 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089839
|
Invitrogen™
MA552887 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDKN2C Recombinant Monoclonal antibody specifically detects CDKN2C in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| CDKN2C | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P42773, Q60772 | |
| 1031, 12580, 54238 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5650 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| C77269; CDK6 inhibitor p18; Cdkn2c; CDKN6; cyclin dependent kinase inhibitor 2C; cyclin-dependent inhibitor; cyclin-dependent kinase 4 inhibitor C; Cyclin-dependent kinase 6 inhibitor; cyclin-dependent kinase 6 inhibitor p18; cyclin-dependent kinase inhibitor 2C (p18, inhibits CDK4); INK4c; p18; p18INK4c; p18-INK4c; p18-INK6 | |
| A synthesized peptide derived from human CDKN2C (1-110AA). | |
| Primary | |
| -20°C | |
| CDKN2C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title