missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDKN2C Recombinant Rabbit Monoclonal Antibody (23GB5650), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552887
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the INK4 family of cyclin-dependent kinase inhibitors. This protein has been shown to interact with CDK4 or CDK6, and prevent the activation of the CDK kinases, thus function as a cell growth regulator that controls cell cycle G1 progression. Ectopic expression of this gene was shown to suppress the growth of human cells in a manner that appears to correlate with the presence of a wild-type RB1 function. Studies in the knockout mice suggested the roles of this gene in regulating spermatogenesis, as well as in suppressing tumorigenesis. Two alternatively spliced transcript variants of this gene, which encode an identical protein, have been reported.
Specifications
| CDKN2C | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CDKN2C | |
| C77269; CDK6 inhibitor p18; Cdkn2c; CDKN6; cyclin dependent kinase inhibitor 2C; cyclin-dependent inhibitor; cyclin-dependent kinase 4 inhibitor C; Cyclin-dependent kinase 6 inhibitor; cyclin-dependent kinase 6 inhibitor p18; cyclin-dependent kinase inhibitor 2C (p18, inhibits CDK4); INK4c; p18; p18INK4c; p18-INK4c; p18-INK6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1031, 12580, 54238 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5650 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P42773, Q60772 | |
| CDKN2C | |
| A synthesized peptide derived from human CDKN2C (1-110AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction