missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdk9 Rabbit anti-Human, Mouse, Rat, Clone: 1C4E3, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £395.00
Specifications
| Antigen | Cdk9 |
|---|---|
| Clone | 1C4E3 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation Sequencing |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP sequencing (ChIP-seq) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18351335
|
Novus Biologicals
NBP3-15345-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18348396
|
Novus Biologicals
NBP3-15345-100UL |
100 μg |
£395.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cdk9 Monoclonal antibody specifically detects Cdk9 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP assay SequencingSpecifications
| Cdk9 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation Sequencing | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| C-2k, CDC2L4CTK1, CDC2-related kinase, Cell division cycle 2-like protein kinase 4, Cell division protein kinase 9, cyclin-dependent kinase 9, cyclin-dependent kinase 9 (CDC2-related kinase), EC 2.7.11, EC 2.7.11.22, EC 2.7.11.23, PITALRE, serine/threonine protein kinase PITALRE, Serine/threonine-protein kinase PITALRE, TAK | |
| A synthetic peptide corresponding to a sequence within amino acids 273-372 of human Cdk9 (P50750). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 1C4E3 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP sequencing (ChIP-seq) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Signal Transduction, Virology Bacteria and Parasites | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 1025 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title