missing translation for 'onlineSavingsMsg'
Learn More

Cdk9 Rabbit anti-Human, Mouse, Rat, Clone: 1C4E3, Novus Biologicals™

Product Code. 18348396 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348396 100 μg 100µL
18351335 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18348396 Supplier Novus Biologicals Supplier No. NBP315345100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Cdk9 Monoclonal antibody specifically detects Cdk9 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP assay Sequencing
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdk9
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP sequencing (ChIP-seq)
Classification Monoclonal
Clone 1C4E3
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation Sequencing
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias C-2k, CDC2L4CTK1, CDC2-related kinase, Cell division cycle 2-like protein kinase 4, Cell division protein kinase 9, cyclin-dependent kinase 9, cyclin-dependent kinase 9 (CDC2-related kinase), EC 2.7.11, EC 2.7.11.22, EC 2.7.11.23, PITALRE, serine/threonine protein kinase PITALRE, Serine/threonine-protein kinase PITALRE, TAK
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 273-372 of human Cdk9 (P50750).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Signal Transduction, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 1025
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.