missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK5RAP2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£153.00 - £359.00
Specifications
| Antigen | CDK5RAP2 |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18682631
|
Novus Biologicals
NBP2-92238-0.02ml |
0.02 mL |
£153.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18686100
|
Novus Biologicals
NBP2-92238-0.1ml |
0.1 mL |
£359.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDK5RAP2 Polyclonal antibody specifically detects CDK5RAP2 in Human samples. It is validated for Western BlotSpecifications
| CDK5RAP2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Cellular Markers, Chromatin Research | |
| PBS with 50% glycerol, pH7.3. | |
| 55755 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| C48, CDK5 activator-binding protein C48, CDK5 regulatory subunit associated protein 2, CDK5 regulatory subunit-associated protein 2, centrosomal protein 215 kDa, Centrosome-associated protein 215, Cep215, DKFZp686B1070, DKFZp686D1070, FLJ10867, KIAA1633microcephaly, primary autosomal recessive 3, MCPH3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1744-1893 of human CDK5RAP2 (NP_060719.4). GQRLLAEMDIQTQEAPSSTSQELGTKGPHPAPLSKFVSSVSTAKLTLEEAYRRLKLLWRVSLPEDGQCPLHCEQIGEMKAEVTKLHKKLFEQEKKLQNTMKLLQLSKRQEKVIFDQLVVTHKILRKARGNLELRPGGAHPGTCSPSRPGS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit