missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK5RAP2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92238-0.1ml
This item is not returnable.
View return policy
Description
CDK5RAP2 Polyclonal antibody specifically detects CDK5RAP2 in Human samples. It is validated for Western Blot
Specifications
| CDK5RAP2 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| C48, CDK5 activator-binding protein C48, CDK5 regulatory subunit associated protein 2, CDK5 regulatory subunit-associated protein 2, centrosomal protein 215 kDa, Centrosome-associated protein 215, Cep215, DKFZp686B1070, DKFZp686D1070, FLJ10867, KIAA1633microcephaly, primary autosomal recessive 3, MCPH3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1744-1893 of human CDK5RAP2 (NP_060719.4). GQRLLAEMDIQTQEAPSSTSQELGTKGPHPAPLSKFVSSVSTAKLTLEEAYRRLKLLWRVSLPEDGQCPLHCEQIGEMKAEVTKLHKKLFEQEKKLQNTMKLLQLSKRQEKVIFDQLVVTHKILRKARGNLELRPGGAHPGTCSPSRPGS | |
| 0.1 mL | |
| Cell Cycle and Replication, Cellular Markers, Chromatin Research | |
| 55755 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction