missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDH10 Recombinant Rabbit Monoclonal Antibody (23GB1600), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | CDH10 |
|---|---|
| Clone | 23GB1600 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089353
![]() |
Invitrogen™
MA552496 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088082
|
Invitrogen™
MA552497 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDH10 Recombinant Monoclonal antibody specifically detects CDH10 in Human, Mouse samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| CDH10 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P70408, Q9Y6N8 | |
| 1008, 320873 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1600 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A830016G23Rik; C030003B10Rik; C030011H18Rik; Cadherin; cadherin 10; cadherin 10 type 2; cadherin 10, type 2 (T2-cadherin); cadherin-10; Cdh10; T2-cadherin | |
| A synthesized peptide derived from human Cadherin 10 (750-788AA). | |
| Primary | |
| -20°C | |
| Cdh10 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title