missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDH10 Recombinant Rabbit Monoclonal Antibody (23GB1600), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552497
This item is not returnable.
View return policy
Description
CDH10 is a type II classical cadherin from the cadherin superfamily, integral membrane proteins that mediate calcium-dependent cell-cell adhesion. Mature cadherin proteins are composed of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small, highly conserved C-terminal cytoplasmic domain. The extracellular domain consists of 5 subdomains, each containing a cadherin motif, and appears to determine the specificity of the protein's homophilic cell adhesion activity. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. This particular cadherin is predominantly expressed in brain and is putatively involved in synaptic adhesions, axon outgrowth and guidance.
Specifications
| CDH10 | |
| Recombinant Monoclonal | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P70408, Q9Y6N8 | |
| Cdh10 | |
| A synthesized peptide derived from human Cadherin 10 (750-788AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1600 | |
| Unconjugated | |
| Cdh10 | |
| A830016G23Rik; C030003B10Rik; C030011H18Rik; Cadherin; cadherin 10; cadherin 10 type 2; cadherin 10, type 2 (T2-cadherin); cadherin-10; Cdh10; T2-cadherin | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1008, 320873 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction