missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDCA7L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£167.00 - £310.00
Specifications
| Antigen | CDCA7L |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2-5 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18188111
|
Novus Biologicals
NBP2-24472SS |
0.025 mg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18116195
|
Novus Biologicals
NBP2-24472 |
0.1 mg |
£310.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDCA7L Polyclonal specifically detects CDCA7L in Human, Primate samples. It is validated for Western Blot.Specifications
| CDCA7L | |
| Western Blot 2-5 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Human, Primate | |
| cell division cycle associated 7-like, DKFZp762L0311, JPO2cell division cycle-associated 7-like protein, Protein JPO2, R1HR1, RAM2, Transcription factor RAM2 | |
| CDCA7L | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 94% homologous in monkey, 89% homologous in mouse, rat and horse, and 84% homologous in cow. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 55536 | |
| A portion of amino acid 170 to 220 of human CDCA7L/RAM2 was used as immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title