missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDCA7L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24472
This item is not returnable.
View return policy
Description
CDCA7L Polyclonal specifically detects CDCA7L in Human, Primate samples. It is validated for Western Blot.
Specifications
| CDCA7L | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| CDCA7L | |
| A portion of amino acid 170 to 220 of human CDCA7L/RAM2 was used as immunogen. | |
| 0.1 mg | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 94% homologous in monkey, 89% homologous in mouse, rat and horse, and 84% homologous in cow. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 2-5 ug/ml | |
| cell division cycle associated 7-like, DKFZp762L0311, JPO2cell division cycle-associated 7-like protein, Protein JPO2, R1HR1, RAM2, Transcription factor RAM2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 55536 | |
| Human, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction