missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDCA3 Recombinant Rabbit Monoclonal Antibody (23GB5045), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£162.00 - £387.00

Specifications

Antigen CDCA3
Clone 23GB5045
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30088376
GREENER_CHOICE
View Documents
Invitrogen™
MA552772
100 μL
£387.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30088980
View Documents
Invitrogen™
MA552773
20 μL
£162.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CDCA3 Recombinant Monoclonal antibody specifically detects CDCA3 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

CDCA3
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human, Mouse
Q99618, Q99M54
14793, 83461
IgG
Affinity chromatography
Antibody
23GB5045
Recombinant Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
2410005A12Rik; C8; CDCA 3; CDCA3; cell division cycle associated 3; cell division cycle-associated protein 3; gene rich cluster, C8; gene-rich cluster protein C8; GRCC 8; GRCC8; MGC2577; TOME 1; Tome1; TOME-1; trigger of mitotic entry 1; trigger of mitotic entry protein 1
A synthesized peptide derived from human CDCA3 (1-42AA).
Primary
-20°C
CDCA3
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.