missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDCA3 Recombinant Rabbit Monoclonal Antibody (23GB5045), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552773
This item is not returnable.
View return policy
Description
F-box-like protein which is required for entry into mitosis. Acts by participating in E3 ligase complexes that mediate the ubiquitination and degradation of WEE1 kinase at G2/M phase. [UniProt].
Specifications
| CDCA3 | |
| Recombinant Monoclonal | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q99618, Q99M54 | |
| CDCA3 | |
| A synthesized peptide derived from human CDCA3 (1-42AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5045 | |
| Unconjugated | |
| CDCA3 | |
| 2410005A12Rik; C8; CDCA 3; CDCA3; cell division cycle associated 3; cell division cycle-associated protein 3; gene rich cluster, C8; gene-rich cluster protein C8; GRCC 8; GRCC8; MGC2577; TOME 1; Tome1; TOME-1; trigger of mitotic entry 1; trigger of mitotic entry protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14793, 83461 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction