missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC7 Rabbit anti-Human, Mouse, Rat, Clone: S01-4G6, Novus Biologicals™
Rabbit Monoclonal Antibody
£232.00 - £469.00
Specifications
| Antigen | CDC7 |
|---|---|
| Clone | S01-4G6 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18360254
|
Novus Biologicals
NBP3-14917-25UL |
25 μg |
£232.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18346235
|
Novus Biologicals
NBP3-14917-100UL |
100 μg |
£469.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDC7 Monoclonal antibody specifically detects CDC7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
| CDC7 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| 8317 | |
| IgG | |
| Affinity-purified |
| S01-4G6 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CDC7 (cell division cycle 7, S. cerevisiae, homolog)-like 1, CDC7 cell division cycle 7 (S. cerevisiae), CDC7L1MGC117361, CDC7-related kinase, cell division cycle 7 (S. cerevisiae), cell division cycle 7 homolog (S. cerevisiae), cell division cycle 7-like protein 1, cell division cycle 7-related protein kinase, EC 2.7.11.1, HsCdc7, Hsk1, huCdc7, MGC126237, MGC126238 | |
| Recombinant protein of human CDC7 (Uniprot#: O00311) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title