missing translation for 'onlineSavingsMsg'
Learn More

CDC7 Rabbit anti-Human, Mouse, Rat, Clone: S01-4G6, Novus Biologicals™

Product Code. 18346235 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346235 100 μg 100µL
18360254 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18346235 Supplier Novus Biologicals Supplier No. NBP314917100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CDC7 Monoclonal antibody specifically detects CDC7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC7
Applications Western Blot, ELISA
Classification Monoclonal
Clone S01-4G6
Conjugate Unconjugated
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias CDC7 (cell division cycle 7, S. cerevisiae, homolog)-like 1, CDC7 cell division cycle 7 (S. cerevisiae), CDC7L1MGC117361, CDC7-related kinase, cell division cycle 7 (S. cerevisiae), cell division cycle 7 homolog (S. cerevisiae), cell division cycle 7-like protein 1, cell division cycle 7-related protein kinase, EC 2.7.11.1, HsCdc7, Hsk1, huCdc7, MGC126237, MGC126238
Host Species Rabbit
Immunogen Recombinant protein of human CDC7 (Uniprot#: O00311)
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 8317
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.