missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC42EP5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£377.00
Specifications
| Antigen | CDC42EP5 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
CDC42EP5 Polyclonal specifically detects CDC42EP5 in Human samples. It is validated for Western Blot.Specifications
| CDC42EP5 | |
| Polyclonal | |
| Rabbit | |
| Binder of Rho GTPases 3, BORG3, CDC42 effector protein (Rho GTPase binding) 5,2010007O02Rik, cdc42 effector protein 5,1700027J19Rik, CEP5MGC21945, MGC71153 | |
| CDC42EP5 | |
| IgG | |
| 15 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 148170 | |
| Synthetic peptides corresponding to CDC42EP5(CDC42 effector protein (Rho GTPase binding) 5) The peptide sequence was selected form the middle region of CDC42EP5. Peptide sequence HVGRGGDAFGDTSFLSRHGGGPPPEPRAPPAGAPRSPPPPAVPQSAAPSP. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title