missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC42EP5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-70491
This item is not returnable.
View return policy
Description
CDC42EP5 Polyclonal specifically detects CDC42EP5 in Human samples. It is validated for Western Blot.
Specifications
| CDC42EP5 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CDC42EP5 | |
| Synthetic peptides corresponding to CDC42EP5(CDC42 effector protein (Rho GTPase binding) 5) The peptide sequence was selected form the middle region of CDC42EP5. Peptide sequence HVGRGGDAFGDTSFLSRHGGGPPPEPRAPPAGAPRSPPPPAVPQSAAPSP. | |
| Affinity purified | |
| RUO | |
| 148170 | |
| Human, Mouse, Rat, Bovine, Canine | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Binder of Rho GTPases 3, BORG3, CDC42 effector protein (Rho GTPase binding) 5,2010007O02Rik, cdc42 effector protein 5,1700027J19Rik, CEP5MGC21945, MGC71153 | |
| Rabbit | |
| 15 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction