missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC37 [p Ser13] Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33562-20ul
This item is not returnable.
View return policy
Description
CDC37 Monoclonal antibody specifically detects CDC37 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| CDC37 | |
| Monoclonal | |
| Western Blot 1:500 - 1:2000, ELISA Recommended starting concentration is 1 μg/mL | |
| CDC37 (cell division cycle 37, S. cerevisiae, homolog), CDC37 cell division cycle 37 homolog, CDC37 cell division cycle 37 homolog (S. cerevisiae), CDC37A, cell division cycle 37 homolog (S. cerevisiae), Hsp90 chaperone protein kinase-targeting subunit, hsp90 co-chaperone Cdc37, P50CDC37 | |
| A synthetic phosphorylated peptide around S13 of human CDC37 (Q16543).,, Sequence:, EVSDD | |
| 20 μL | |
| Cell Cycle and Replication, Membrane Trafficking and Chaperones, Phospho Specific | |
| 11140 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction