missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC37 [p Ser13] Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£161.00 - £383.00
Specifications
| Antigen | CDC37 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231343
|
Novus Biologicals
NBP3-33562-20ul |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228109
|
Novus Biologicals
NBP3-33562-100ul |
100 μL |
£383.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDC37 Monoclonal antibody specifically detects CDC37 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| CDC37 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Membrane Trafficking and Chaperones, Phospho Specific | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 11140 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CDC37 (cell division cycle 37, S. cerevisiae, homolog), CDC37 cell division cycle 37 homolog, CDC37 cell division cycle 37 homolog (S. cerevisiae), CDC37A, cell division cycle 37 homolog (S. cerevisiae), Hsp90 chaperone protein kinase-targeting subunit, hsp90 co-chaperone Cdc37, P50CDC37 | |
| A synthetic phosphorylated peptide around S13 of human CDC37 (Q16543).,, Sequence:, EVSDD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title