missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDC25C Recombinant Rabbit Monoclonal Antibody (23GB3430), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | CDC25C |
|---|---|
| Clone | 23GB3430 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30088852
|
Invitrogen™
MA553378 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089855
|
Invitrogen™
MA553379 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDC25C Recombinant Monoclonal antibody specifically detects CDC25C in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| CDC25C | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P30307 | |
| 995 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3430 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| CDC25; CDC25C; Cdc25m1; cell division cycle 25C; dual specificity phosphatase CDC25C; EC 3.1.3.48; mitosis inducer CDC25; M-phase inducer phosphatase 3; MPIP3; OTTHUMP00000159490; OTTHUMP00000224012; OTTHUMP00000224013; OTTHUMP00000224016; phosphotyrosine phosphatase; PPP1R60; protein phosphatase 1, regulatory subunit 60 | |
| A synthetic peptide of human Cdc25C (11-63AA). | |
| Primary | |
| -20°C | |
| Cdc25c |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title