missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDC25C Recombinant Rabbit Monoclonal Antibody (23GB3430), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553378
This item is not returnable.
View return policy
Description
M-phase inducer phosphatase 3is an enzyme that in humans is encoded by theCDC25Cgene. This gene is highly conserved during evolution and it plays a key role in the regulation of cell division. The encoded protein is a tyrosine phosphatase and belongs to the Cdc25 phosphatase family. It directs dephosphorylation of cyclin B-bound CDC2 (CDK1) and triggers entry into mitosis. Also, it is thought to suppress p53-induced growth arrest. Multiple alternatively spliced transcript variants of this gene have been described, however, the full-length nature of many of them is not known.
Specifications
| CDC25C | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Cdc25c | |
| CDC25; CDC25C; Cdc25m1; cell division cycle 25C; dual specificity phosphatase CDC25C; EC 3.1.3.48; mitosis inducer CDC25; M-phase inducer phosphatase 3; MPIP3; OTTHUMP00000159490; OTTHUMP00000224012; OTTHUMP00000224013; OTTHUMP00000224016; phosphotyrosine phosphatase; PPP1R60; protein phosphatase 1, regulatory subunit 60 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 995 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3430 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P30307 | |
| Cdc25c | |
| A synthetic peptide of human Cdc25C (11-63AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction