missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cdc23 Recombinant Rabbit Monoclonal Antibody (23GB3420), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | Cdc23 |
|---|---|
| Clone | 23GB3420 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087779
![]() |
Invitrogen™
MA553384 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089036
|
Invitrogen™
MA553385 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cdc23 Recombinant Monoclonal antibody specifically detects Cdc23 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| Cdc23 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q8BGZ4, Q9UJX2 | |
| 291689, 52563, 8697 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3420 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 6030435O18; ANAPC8; anaphase promoting complex subunit 8; anaphase-promoting complex subunit 8; APC8; CDC23; CDC23 (cell division cycle 23, yeast, homolog); CDC23 cell division cycle 23; cell division cycle 23; cell division cycle 23 homolog; cell division cycle protein 23; cell division cycle protein 23 homolog; CUT23; cyclosome subunit 8; D18Ertd243e; zgc:56013 | |
| A synthetic peptide of human Cdc23/APC8 (510-575AA). | |
| Primary | |
| -20°C | |
| Cdc23 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title