missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cdc23 Recombinant Rabbit Monoclonal Antibody (23GB3420), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553385
This item is not returnable.
View return policy
Description
Ubiquitination is an important mechanism through which three classes of enzymes act in concert to target short-lived or abnormal proteins for destruction. The three classes of enzymes involved in ubiquitination are the ubiquitin-activating enzymes (E1s), the ubiquitin-conjugating enzymes (E2s) and the ubiquitin-protein ligases (E3s). Ubr2 (Ubiquitin-protein ligase E3-alpha-2), also known as N-recognin-2, is a 1755 amino acid protein that contains one UBR-type zinc finger and one RING-type zinc finger. Participating in protein modification events within the N-end rule pathway, Ubr2 functions as an E3 ubiquitin-protein ligase that recognizes and binds proteins that contain destabilizing N-terminal residues, thereby leading to their ubiquitination and subsequent degradation. Mice lacking Ubr2 are infertile due to defects in male meiosis.
Specifications
| Cdc23 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Cdc23 | |
| 6030435O18; ANAPC8; anaphase promoting complex subunit 8; anaphase-promoting complex subunit 8; APC8; CDC23; CDC23 (cell division cycle 23, yeast, homolog); CDC23 cell division cycle 23; cell division cycle 23; cell division cycle 23 homolog; cell division cycle protein 23; cell division cycle protein 23 homolog; CUT23; cyclosome subunit 8; D18Ertd243e; zgc:56013 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 291689, 52563, 8697 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3420 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BGZ4, Q9UJX2 | |
| Cdc23 | |
| A synthetic peptide of human Cdc23/APC8 (510-575AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction