missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD99 Antibody (MIC2/1495R), Novus Biologicals™
Rabbit Monoclonal Antibody
£227.00 - £432.00
Specifications
| Antigen | CD99 |
|---|---|
| Clone | MIC2/1495R |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 2 - 4ug/million cells, Immunocytochemistry/Immunofluorescence 2 - 4 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18257190
|
Novus Biologicals
NBP2-53368-20UG |
20 μg |
£227.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18244290
|
Novus Biologicals
NBP2-53368-100UG |
100 μg |
£432.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD99 Monoclonal specifically detects CD99 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CD99 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Immunology | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 4267 | |
| Recombinant human full-length MIC2 protein | |
| Primary | |
| Store at 4C. |
| MIC2/1495R | |
| Flow Cytometry 2 - 4ug/million cells, Immunocytochemistry/Immunofluorescence 2 - 4 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 12E7, antigen identified by monoclonal antibodies 12E7, F21 and O13, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2 | |
| CD99 | |
| IgG κ | |
| Protein A purified | |
| 29 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title