missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD99 Antibody (MIC2/1495R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-53368-20UG
This item is not returnable.
View return policy
Description
CD99 Monoclonal specifically detects CD99 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CD99 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 2 - 4ug/million cells, Immunocytochemistry/Immunofluorescence 2 - 4 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml | |
| 12E7, antigen identified by monoclonal antibodies 12E7, F21 and O13, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2 | |
| Rabbit | |
| 29 kDa | |
| 20 μg | |
| Immunology | |
| 4267 | |
| Store at 4C. | |
| IgG κ |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| MIC2/1495R | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| CD99 | |
| Recombinant human full-length MIC2 protein | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction