missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD97 Antibody (2Q42) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£161.00 - £426.00
Specifications
| Antigen | CD97 |
|---|---|
| Clone | 2Q42 |
| Concentration | LYOPH |
| Applications | CyTOF, Flow Cytometry |
| Classification | Monoclonal |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18133464
|
Novus Biologicals
NBP2-22000-0.025MG |
0.025 mg |
£161.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18106741
|
Novus Biologicals
NBP2-22000 |
0.1 mg |
£426.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
CD97 Monoclonal specifically detects CD97 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Spezifikation
| CD97 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1 | |
| IgG1 | |
| Protein G purified |
| 2Q42 | |
| CyTOF, Flow Cytometry | |
| Unconjugated | |
| Rat | |
| GPCR, Immunology | |
| AAH06676 | |
| 976 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts