missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD97 Antibody (2Q42) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-22000
This item is not returnable.
View return policy
Description
CD97 Monoclonal specifically detects CD97 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| CD97 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 1:10-1:50, CyTOF-ready | |
| AAH06676 | |
| ADGRE5 | |
| Mouse recombinant CD97 | |
| 0.1 mg | |
| GPCR, Immunology | |
| 976 | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 2Q42 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative | |
| CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reconstitute with sterilized PBS to a final concentration of 0.2 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction