missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD94 Mouse anti-Human, BB700, Clone: HP-3D9, BD Horizon™
Mouse Monoclonal Antibody
£305.00 - £541.00
Specifications
| Antigen | CD94 |
|---|---|
| Clone | HP-3D9 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Blue 700 (BB700) |
Description
CD94 Monoclonal antibody specifically detects CD94 in Human samples. It is validated for Flow Cytometry.Specifications
| CD94 | |
| Flow Cytometry | |
| Brilliant Blue 700 (BB700) | |
| Mouse | |
| Human | |
| Primary |
| HP-3D9 | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG1 κ | |
| Refrigerate (2 to 8°C) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title