missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD94 Mouse anti-Human, BB700, Clone: HP-3D9, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566535
This item is not returnable.
View return policy
Description
CD94 Monoclonal antibody specifically detects CD94 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD94 | |
| Monoclonal | |
| Brilliant Blue 700 (BB700) | |
| 25 Tests | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ |
| Flow Cytometry | |
| HP-3D9 | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction