missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD94 Antibody (DX22) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64104-0.025MG
This item is not returnable.
View return policy
Description
CD94 Monoclonal specifically detects CD94 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD94 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 1:50-1:100 | |
| Q13241 | |
| KLRD1 | |
| Natural killer cell line | |
| 0.025 mg | |
| Innate Immunity | |
| 3824 | |
| Human | |
| Purified |
| Flow Cytometry | |
| DX22 | |
| Unconjugated | |
| PBS (pH 7.4) with 0.09% Sodium Azide | |
| CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KP43, NK cell receptor | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-64104 is specific for human CD94, also known as KLRD1 (Killer cell lectin-like receptor, subfamily D, member 1). CD94 is expressed on natural killer (NK) cells and a subset of T lymphocytes. CD94 is found to associate with NKG2 to form a heterodimer which is involved in the inhibition of cell mediated cytotoxicity against cells bearing appropriate MHC class I allotypes. Clone DX22 is reported to inhibit the binding of CD94 to HLA-E(2) and HLA-G(4). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction