missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD94 Antibody (DX22) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£170.00 - £426.00
Specifications
| Antigen | CD94 |
|---|---|
| Clone | DX22 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 1:50-1:100 |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18013116
|
Novus Biologicals
NB100-64104-0.025MG |
0.025 mg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18061564
|
Novus Biologicals
NB100-64104 |
0.1 mg |
£426.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD94 Monoclonal specifically detects CD94 in Human samples. It is validated for Flow Cytometry.Specifications
| CD94 | |
| 1.0 mg/mL | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Innate Immunity | |
| Q13241 | |
| 3824 | |
| Natural killer cell line | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| DX22 | |
| Flow Cytometry 1:50-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KP43, NK cell receptor | |
| KLRD1 | |
| IgG1 | |
| Protein G purified | |
| NB100-64104 is specific for human CD94, also known as KLRD1 (Killer cell lectin-like receptor, subfamily D, member 1). CD94 is expressed on natural killer (NK) cells and a subset of T lymphocytes. CD94 is found to associate with NKG2 to form a heterodimer which is involved in the inhibition of cell mediated cytotoxicity against cells bearing appropriate MHC class I allotypes. Clone DX22 is reported to inhibit the binding of CD94 to HLA-E(2) and HLA-G(4). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title