missing translation for 'onlineSavingsMsg'
Learn More

CD94 Antibody (3G2), Novus Biologicals™

Mouse Monoclonal Antibody

£173.00 - £435.00

Specifications

Antigen CD94
Clone 3G2
Dilution Western Blot 1:500 - 1:2000, ELISA 1:10000
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18645215
View Documents
Novus Biologicals
NBP2-37631-0.025ml
0.025 mL
£173.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18182908
View Documents
Novus Biologicals
NBP2-37631
0.1 mL
£435.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD94 Monoclonal specifically detects CD94 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

CD94
Western Blot 1:500 - 1:2000, ELISA 1:10000
Monoclonal
Ascites
RUO
Human
CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KP43, NK cell receptor
KLRD1
IgG1
Unpurified
3G2
Western Blot, ELISA
Unconjugated
Mouse
Innate Immunity
Ascites with 0.03% Sodium Azide
3824
Purified recombinant fragment of human CD94 expressed in E. Coli.
Primary
21 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.