missing translation for 'onlineSavingsMsg'
Learn More

CD94 Antibody (3G2), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-37631

 View more versions of this product

Product Code. 18182908

  • £435.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD94 Monoclonal specifically detects CD94 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD94
Monoclonal
Unconjugated
Ascites with 0.03% Sodium Azide
KLRD1
Purified recombinant fragment of human CD94 expressed in E. Coli.
Unpurified
RUO
Primary
Human
Ascites
Western Blot, ELISA
3G2
Western Blot 1:500 - 1:2000, ELISA 1:10000
CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KP43, NK cell receptor
Mouse
21 kDa
0.1 mL
Innate Immunity
3824
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only