missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD94 Antibody (3G2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37631-0.025ml
This item is not returnable.
View return policy
Description
CD94 Monoclonal specifically detects CD94 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| CD94 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| KLRD1 | |
| Purified recombinant fragment of human CD94 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
| Western Blot, ELISA | |
| 3G2 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KP43, NK cell receptor | |
| Mouse | |
| 21 kDa | |
| 0.025 mL | |
| Innate Immunity | |
| 3824 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction